5-Ethyluracil, 98+%, Thermo Scientific Chemicals
5-Ethyluracil, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

5-Ethyluracil, 98+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
Catalog number L10861.MD
also known as L10861-MD
Price (USD)/ Each
-
Quantity:
250 mg
Request bulk or custom format
Chemical Identifiers
CAS4212-49-1
IUPAC Name5-ethyl-1,2,3,4-tetrahydropyrimidine-2,4-dione
Molecular FormulaC6H8N2O2
InChI KeyRHIULBJJKFDJPR-UHFFFAOYSA-N
SMILESCCC1=CNC(=O)NC1=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)≥98.0%
Appearance (Color)White
FormCrystals or powder or crystalline powder
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only