Thermo Scientific Chemicals

4-Nitrophenyl palmitate, 98+%, Thermo Scientific Chemicals

Catalog number: L10896.06
5 g, Each
Thermo Scientific Chemicals

4-Nitrophenyl palmitate, 98+%, Thermo Scientific Chemicals

Catalog number: L10896.06
5 g, Each
Quantity
Catalog number: L10896.06
also known as L10896-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1492-30-4
IUPAC Name
4-nitrophenyl hexadecanoate
Molecular Formula
C22H35NO4
InChI Key
LVZSQWIWCANHPF-UHFFFAOYSA-N
SMILES
CCCCCCCCCCCCCCCC(=O)OC1=CC=C(C=C1)[N+]([O-])=O
Assay (GC)
≥98.0%
Melting Point
61.0-67.0?C
Appearance (Color)
White to pale cream
Form
Crystals or powder or crystalline powder
Identification (FTIR)
Conforms

Description

The activities of free and immobilized PPL were determined by spectrophotometric assay using 4-nitrophenyl palmitate (pNPP) as substrate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
The activities of free and immobilized PPL were determined by spectrophotometric assay using 4-nitrophenyl palmitate (pNPP) as substrate.

Solubility
Insoluble in water. Soluble in CHCl3.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents. Store at -20°C
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text