Thermo Scientific Chemicals

4-Vinylphenol, min 10% soln. in propylene glycol, Thermo Scientific Chemicals

Catalog number: L10902.03
1 g, Each
Thermo Scientific Chemicals

4-Vinylphenol, min 10% soln. in propylene glycol, Thermo Scientific Chemicals

Catalog number: L10902.03
1 g, Each
Quantity
Catalog number: L10902.03
also known as L10902-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
2628-17-3
IUPAC Name
4-ethenylphenol
Molecular Formula
C8H8O
InChI Key
FUGYGGDSWSUORM-UHFFFAOYSA-N
SMILES
OC1=CC=C(C=C)C=C1
Form
Liquid
Appearance (Color)
Clear colorless to yellow or orange
Assay from Supplier's CofA
≥9.0 to ≤15.0%
Refractive Index
1.4420-1.4520 @ 20?C

Description

4-Vinylphenol acts as a monomer and used in the preparation of poly-4-vinylphenol polymer, which is used in electronics as a dielectric layer in organic transistors in organic thin-film-transistor liquid-crystal displays (TFT LCD). It is a phenolic compound and presents in wine and beer.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Vinylphenol acts as a monomer and used in the preparation of poly-4-vinylphenol polymer, which is used in electronics as a dielectric layer in organic transistors in organic thin-film-transistor liquid-crystal displays (TFT LCD). It is a phenolic compound and presents in wine and beer.

Solubility
Slightly miscible with water.

Notes
Hygroscopic. Moisture and heat sensitive. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text