Specifications
Chemical Name or MaterialEthyl 3-isothiocyanatopropionate
CAS17126-62-4
Health Hazard 1H302+H312-H314-H331-H335
Health Hazard 2GHS H Statement
H331-H314-H318-H302-H312
Toxic if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.
Health Hazard 3P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
View more