Thermo Scientific Chemicals

4-Benzyl-3-thiosemicarbazide, 98+%, Thermo Scientific Chemicals

Catalog number: L10920.03
1 g, Each
Thermo Scientific Chemicals

4-Benzyl-3-thiosemicarbazide, 98+%, Thermo Scientific Chemicals

Catalog number: L10920.03
1 g, Each
Quantity
Catalog number: L10920.03
also known as L10920-03
Price (USD)
Quantity
-
Have Questions?

Description

4-Benzyl-3-thiosemicarbazide is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Benzyl-3-thiosemicarbazide is used as a pharmaceutical intermediate.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents. Stable under recommended storage conditions.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text