6-Hydroxy-2,3-dihydrobenzo[b]furan-3-one, 97%, Thermo Scientific Chemicals
6-Hydroxy-2,3-dihydrobenzo[b]furan-3-one, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

6-Hydroxy-2,3-dihydrobenzo[b]furan-3-one, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Catalog number L10951.06
also known as L10951-06
Price (USD)/ Each
197.00
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
197.00
Add to cart
6-Hydroxy-2,3-dihydrobenzo[b]furan-3-one, 97%, Thermo Scientific Chemicals
Catalog numberL10951.06
Price (USD)/ Each
197.00
-
Add to cart
Chemical Identifiers
CAS6272-26-0
IUPAC Name6-hydroxy-2,3-dihydro-1-benzofuran-3-one
Molecular FormulaC8H6O3
InChI KeyGBDMODVZBPFQKI-UHFFFAOYSA-N
SMILESOC1=CC=C2C(=O)COC2=C1
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder or fused solid
Assay (HPLC)≥96.0%
Appearance (Color)Yellow to orange to brown
6-Hydroxy-2,3-dihydrobenzo[b]furan-3-one, is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
6-Hydroxy-2,3-dihydrobenzo[b]furan-3-one, is used as a pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents. Stable under recommended storage conditions.
RUO – Research Use Only
C Avitia-Domínguez.; E Sierra-Campos. Inhibition and Biochemical Characterization of Methicillin-Resistant Staphylococcus aureus Shikimate Dehydrogenase: An in Silico and Kinetic Study. Molecules. 2014, 19,(4), 4491-4509.