Thermo Scientific Chemicals

1,2,3,4,5-Pentamethylcyclopentadiene, 94%, Thermo Scientific Chemicals

Catalog number: L10957.14
25 g, Each
Thermo Scientific Chemicals

1,2,3,4,5-Pentamethylcyclopentadiene, 94%, Thermo Scientific Chemicals

Catalog number: L10957.14
25 g, Each
Quantity
Catalog number: L10957.14
also known as L10957-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
4045-44-7
IUPAC Name
1,2,3,4,5-pentamethylcyclopenta-1,3-diene
Molecular Formula
C10H16
InChI Key
WQIQNKQYEUMPBM-UHFFFAOYSA-N
SMILES
CC1C(C)=C(C)C(C)=C1C
Identification (FTIR)
Conforms
Assay (GC)
≥92.0%
Refractive Index
1.4710-1.4780 @ 20?C
Appearance (Color)
Clear colorless to yellow
Form
Liquid

Description

1,2,3,4,5-Pentamethylcyclopentadiene serves as a precursor to the ligand 1,2,3,4,5-pentamethylcyclopentadienyl. It is used as a growth modifier chemical, during metal organic chemical vapor deposition of iron from iron pentacarbonyl. It is used as a ligand in one-pot iridium-catalyzed transformation of alcohols to amides. Further, it acts as an electron mediator in the regeneration process of nicotinamide adenine dinucleotide. In addition to this, it undergoes radical cation catalyzed cycloaddition reaction with allenes to get Diels-Alder product.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,2,3,4,5-Pentamethylcyclopentadiene serves as a precursor to the ligand 1,2,3,4,5-pentamethylcyclopentadienyl. It is used as a growth modifier chemical, during metal organic chemical vapor deposition of iron from iron pentacarbonyl. It is used as a ligand in “one-pot” iridium-catalyzed transformation of alcohols to amides. Further, it acts as an electron mediator in the regeneration process of nicotinamide adenine dinucleotide. In addition to this, it undergoes radical cation catalyzed cycloaddition reaction with allenes to get Diels-Alder product.

Solubility
Miscible with methanol. dichloromethane and ethyl acetate. Slightly miscible with water.

Notes
Light sensitive. Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text