2,4,6-Trimethylphenylhydrazine hydrochloride, 97%, Thermo Scientific Chemicals
2,4,6-Trimethylphenylhydrazine hydrochloride, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2,4,6-Trimethylphenylhydrazine hydrochloride, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number L10958.03
also known as L10958-03
Price (USD)/ Each
104.00
-
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)/ Each
104.00
Add to cart
2,4,6-Trimethylphenylhydrazine hydrochloride, 97%, Thermo Scientific Chemicals
Catalog numberL10958.03
Price (USD)/ Each
104.00
-
Add to cart
Chemical Identifiers
CAS76195-82-9
IUPAC Name(2,4,6-trimethylphenyl)hydrazine hydrochloride
Molecular FormulaC9H15ClN2
InChI KeyXFGPCULNJGZRTH-UHFFFAOYSA-N
SMILESCl.CC1=CC(C)=C(NN)C(C)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder or lumps or flakes
Assay (Titration ex Chloride)≥96.0 to ≤104.0%
Appearance (Color)White to cream to orange to pale brown
Identification (FTIR)Conforms
2,4,6-Trimethylphenylhydrazine hydrochloride is used as pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,4,6-Trimethylphenylhydrazine hydrochloride is used as pharmaceutical intermediates.

Solubility
Soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents.
RUO – Research Use Only

General References:

  1. Robert B. Carlin.; Mead S. Moores. The Fischer Reaction of Cyclohexanone Mesitylhydrazone. Evidence of a 1,4-Methyl Migration. J. Am. Chem. Soc. 1962, 84 (21), 4107-4112.
  2. Rudolph A. Abramovitch.; Joseph M. Beckert.; Harry H. Gibson, Jr.; Anthony Belcher.; Greg Hundt.; Tommy Sierra.; Santiago Olivella.; William T. Pennington.; Albert Solé. The 1,2,4-Triazolyl Cation: Thermolytic and Photolytic Studies. J. Org. Chem. 2001, 66 (4), 1242-1251.