Thiophene-2-carboxaldoxime, 98+%, Thermo Scientific Chemicals
Thiophene-2-carboxaldoxime, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Thiophene-2-carboxaldoxime, 98+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number L10961.06
also known as L10961-06
Price (USD)/ Each
49.70
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
49.70
Add to cart
Thiophene-2-carboxaldoxime, 98+%, Thermo Scientific Chemicals
Catalog numberL10961.06
Price (USD)/ Each
49.70
-
Add to cart
Chemical Identifiers
CAS29683-84-9
SpecificationsSpecification SheetSpecification Sheet
Melting Point132-138?C
FormCrystals or crystalline powder
Appearance (Color)white to pale cream
Assay (HPLC)≥98.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only