Thermo Scientific Chemicals

1-Octadecene, tech. 90%, Thermo Scientific Chemicals

Catalog number: L11004.AU
1000 mL, Each
Thermo Scientific Chemicals

1-Octadecene, tech. 90%, Thermo Scientific Chemicals

Catalog number: L11004.AU
1000 mL, Each
Quantity
Catalog number: L11004.AU
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
112-88-9
IUPAC Name
octadec-1-ene
Molecular Formula
C18H36
InChI Key
CCCMONHAUSKTEQ-UHFFFAOYSA-N
SMILES
CCCCCCCCCCCCCCCCC=C
Appearance (Color)
Clear colorless to pale yellow
Form
Liquid
Assay (GC)
≥88.0% (UK-sourced material)
Assay from Supplier's CofA
≥88.0% (US-sourced material)
Identification (FTIR)
Conforms (UK-sourced material)

Description

1-Octadecene is an inexpensive solvent having high boiling point. Due to this property, it replaces the heat transfer fluids such as dowthem A or therminol 66 and used in the synthesis of high quality CdSe quantum dots (nano crystal made up of semiconductor materials). These quantum dots find application in transistors, diode lasers and solar cells. It is also used as a solvent for the synthesis of iron oxide nanospheres.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Octadecene is an inexpensive solvent having high boiling point. Due to this property, it replaces the heat transfer fluids such as dowthem A or therminol 66 and used in the synthesis of high quality CdSe quantum dots (nano crystal made up of semiconductor materials). These quantum dots find application in transistors, diode lasers and solar cells. It is also used as a solvent for the synthesis of iron oxide nanospheres.

Solubility
Miscible with alcohol, acetone and ether. Immiscible with water.

Notes
Avoid reaction with strong Lewis or mineral acids. Free radical initiators should be avoided. Keep container tightly closed in a dry and well-ventilated place.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text