Thermo Scientific Chemicals

2-Bromoethyl phenyl sulfide, 97%, Thermo Scientific Chemicals

Catalog number: L11005.14
25 g, Each
Thermo Scientific Chemicals

2-Bromoethyl phenyl sulfide, 97%, Thermo Scientific Chemicals

Catalog number: L11005.14
25 g, Each
Quantity
Catalog number: L11005.14
also known as L11005-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
4837-01-8
IUPAC Name
[(2-bromoethyl)sulfanyl]benzene
Molecular Formula
C8H9BrS
InChI Key
UEFBOQYLXLEJSM-UHFFFAOYSA-N
SMILES
BrCCSC1=CC=CC=C1
Appearance (Color)
Clear colorless to yellow to green to brown to dark brown or grey
Form
Liquid
Assay (GC)
≥96.0%
Refractive Index
1.6050-1.6100 @ 20?C

Description

2-Bromoethyl phenyl sulfide, is used to make other chemicals as well as pharmaceutical intermediate. It is also used as a application intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Bromoethyl phenyl sulfide, is used to make other chemicals as well as pharmaceutical intermediate. It is also used as a application intermediate.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents. Stable under recommended storage conditions.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text