Thermo Scientific Chemicals

DL-Alanine ethyl ester hydrochloride, 99%, Thermo Scientific Chemicals

Catalog number: L11020.14
25 g, Each
Thermo Scientific Chemicals

DL-Alanine ethyl ester hydrochloride, 99%, Thermo Scientific Chemicals

Catalog number: L11020.14
25 g, Each
Quantity
Catalog number: L11020.14
also known as L11020-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
617-27-6
IUPAC Name
ethyl 2-aminopropanoate hydrochloride
Molecular Formula
C5H12ClNO2
InChI Key
JCXLZWMDXJFOOI-UHFFFAOYNA-N
SMILES
Cl.CCOC(=O)C(C)N
Form
Powder or lumps
Assay (Titration ex Chloride)
≥98.5 to ≤101.5%
Appearance (Color)
White

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text