(R)-(-)-2-Amino-1-propanol is used as a chiral intermediate for the synthesis of pharmaceutical products and agrochemicals. It acts as additives in health care products and animal feed.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
(R)-(-)-2-Amino-1-propanol is used as a chiral intermediate for the synthesis of pharmaceutical products and agrochemicals. It acts as additives in health care products and animal feed.
Solubility
Miscible with water.
Notes
Air sensitive and hygroscopic. Keep away from sources of ignition. Keep the container tightly closed in a dry and well-ventilated place.
RUO – Research Use Only
General References:
- Contini, G.; Gori, P.; Di Giovannantonio, M.; Zema, N.; Turchini, S.; Catone, D.; Prosperi, T.; Palma, A. Interplay between Supramolecularity and Substrate Symmetry in the Dehydrogenation of D-Alaninol on Cu(100) and Cu(110) Surfaces. J. Phys. Chem. C 2013, 117 (20), 10545-10551.
- Gori, P.; Contini, G.; Ronci, F.; Colonna, S.; Zema, N.; Turchini, S.; Catone, D.; Cricenti, A.; Prosperi, T.; Aschi, M.; Palma, A. Self assembly and chirality transfer in D-Alaninol on the Cu(100) surface. Eur. Phys. J. D 2012, 66,136.