Thermo Scientific Chemicals

4-Nitrophenyl isothiocyanate, 97%, Thermo Scientific Chemicals

Catalog number: L11042.18
50 g, Each
Thermo Scientific Chemicals

4-Nitrophenyl isothiocyanate, 97%, Thermo Scientific Chemicals

Catalog number: L11042.18
50 g, Each
Quantity
Catalog number: L11042.18
also known as L11042-18
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
2131-61-5
IUPAC Name
1-isothiocyanato-4-nitrobenzene
Molecular Formula
C7H4N2O2S
InChI Key
NXHSSIGRWJENBH-UHFFFAOYSA-N
SMILES
[O-][N+](=O)C1=CC=C(C=C1)N=C=S
Identification (FTIR)
Conforms
Melting Point (clear melt)
106-112?C
Form
Powder or lumpy solid
Appearance (Color)
Cream to yellow
Assay (GC)
≥96.0%

Description

4-Nitrophenyl isothiocyanate is used as specializing in synthesizing of organic fine chemicals and pharmaceutical intermediates for use in development and production

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Nitrophenyl isothiocyanate is used as specializing in synthesizing of organic fine chemicals and pharmaceutical intermediates for use in development and production

Solubility
It hydrolyzes with water. Soluble in toluene.

Notes
Moisture Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents. Stable under recommended storage conditions.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text