1,1,1,2,2-Pentafluoro-3-iodopropane is used to produce 1,1,1,2,2-pentafluoro-propane by heating. It is an important raw material and intermediate used in organic synthesis, pharmaceuticals and agrochemicals.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
1,1,1,2,2-Pentafluoro-3-iodopropane is used to produce 1,1,1,2,2-pentafluoro-propane by heating. It is an important raw material and intermediate used in organic synthesis, pharmaceuticals and agrochemicals.
Solubility
Insoluble in water.
Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers. It is sensitive to light. Incompatible with strong oxidizing agents.
RUO – Research Use Only
General References:
- RS Dickson.; GD Sutcliffe. Fluorocarbon-Aluminum compounds. III. Reactions of Heptafluoro-3-iodopropane, 1,1,1,2,2-Pentafluoro-3-iodopropane, and Heptafluoro-2-iodopropane with lithium tetrahydroaluminate. Australian Journal of Chemistry. 1972, 25 (4), 761 - 768 .
- Apeng Liang.; Xinjian Li.; Dongfeng Liu.; Jingya Li.; Dapeng Zou.; Yangjie Wu.; Yusheng Wu. The palladium-catalyzed cross-coupling reactions of trifluoroethyl iodide with aryl and heteroaryl boronic acid esters. Chem. Commun. 2012, 48 8273-8275.