Thermo Scientific Chemicals

1,1,1,2,2-Pentafluoro-3-iodopropane, 96%, Thermo Scientific Chemicals

Catalog number: L11047.06
5 g, Each
Thermo Scientific Chemicals

1,1,1,2,2-Pentafluoro-3-iodopropane, 96%, Thermo Scientific Chemicals

Catalog number: L11047.06
5 g, Each
Quantity
Catalog number: L11047.06
also known as L11047-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
354-69-8
IUPAC Name
1,1,1,2,2-pentafluoro-3-iodopropane
Molecular Formula
C3H2F5I
InChI Key
HENALDZJQYAUBN-UHFFFAOYSA-N
SMILES
FC(F)(F)C(F)(F)CI
Appearance (Color)
Clear colorless
Form
Liquid
Identification (FTIR)
Conforms
Assay (GC)
≥95.0%
Refractive Index
1.3700-1.3770 @ 20?C

Description

1,1,1,2,2-Pentafluoro-3-iodopropane is used to produce 1,1,1,2,2-pentafluoro-propane by heating. It is an important raw material and intermediate used in organic synthesis, pharmaceuticals and agrochemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,1,1,2,2-Pentafluoro-3-iodopropane is used to produce 1,1,1,2,2-pentafluoro-propane by heating. It is an important raw material and intermediate used in organic synthesis, pharmaceuticals and agrochemicals.

Solubility
Insoluble in water.

Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers. It is sensitive to light. Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text