Thermo Scientific Chemicals

1,10-Decanedithiol, 95%, Thermo Scientific Chemicals

Catalog number: L11064.06
5 g, Each
Thermo Scientific Chemicals

1,10-Decanedithiol, 95%, Thermo Scientific Chemicals

Catalog number: L11064.06
5 g, Each
Quantity
Catalog number: L11064.06
also known as L11064-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1191-67-9
IUPAC Name
decane-1,10-dithiol
Molecular Formula
C10H22S2
InChI Key
UOQACRNTVQWTFF-UHFFFAOYSA-N
SMILES
SCCCCCCCCCCS
Assay (GC)
≥94.0%
Appearance (Color)
Clear colorless to white
Identification (FTIR)
Conforms
Refractive Index
1.4960-1.5020 @ 20?C
Form
Liquid

Description

1,10-Decanedithiol is an important raw material and intermediate used in organic synthesis, pharmaceuticals and agrochemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,10-Decanedithiol is an important raw material and intermediate used in organic synthesis, pharmaceuticals and agrochemicals.

Solubility
Insoluble in water.

Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers. It is sensitive to air. Store under inert gas.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text