1,10-Decanedithiol is an important raw material and intermediate used in organic synthesis, pharmaceuticals and agrochemicals.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
1,10-Decanedithiol is an important raw material and intermediate used in organic synthesis, pharmaceuticals and agrochemicals.
Solubility
Insoluble in water.
Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers. It is sensitive to air. Store under inert gas.
RUO – Research Use Only
Takuji Ogawa.; Keijiro Kobayashi.; Gou Masuda.; Takuya Takase.; Seisuke Maeda. Electronic conductive characteristics of devices fabricated with 1,10-decanedithiol and gold nanoparticles between 1-μm electrode gaps. Thin Solid Films. 2001, 393 (1-2), 374-378.
E.M.S. Azzam.; A. Bashir, O. Shekhah.; A.R.E. Alawady.; A. Birkner.; Ch. Grunwald.; Ch. Wöll. Fabrication of a surface plasmon resonance biosensor based on gold nanoparticles chemisorbed onto a 1,10-decanedithiol self-assembled monolayer. Thin Solid Films. 2009, 518 (1), 387-391.