2-Cyclopropylethanol, 96%, Thermo Scientific Chemicals
2-Cyclopropylethanol, 96%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-Cyclopropylethanol, 96%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
5 g
Catalog number L11080.MD
also known as L11080-MD
Price (USD)/ Each
48.40
-
Add to cart
Quantity:
250 mg
Request bulk or custom format
Price (USD)/ Each
48.40
Add to cart
2-Cyclopropylethanol, 96%, Thermo Scientific Chemicals
Catalog numberL11080.MD
Price (USD)/ Each
48.40
-
Add to cart
Chemical Identifiers
CAS2566-44-1
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear, colourless to pale yellow
Formliquid
Assay (GC)>95.0%
Refractive Index1.4325 - 1.4385
2-Cyclopropylethanol is used to prepare (2-bromo-ethyl)-cyclopropane by reacting with phosphorus tribromide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Cyclopropylethanol is used to prepare (2-bromo-ethyl)-cyclopropane by reacting with phosphorus tribromide.

Solubility
Miscible with water.

Notes
Incompatible with oxidizing agents.
RUO – Research Use Only

General References:

  1. Møllendal, H.; Frank, D.; de Meijere, A. Structural and conformational properties and intramolecular hydrogen bonding of (methylenecyclopropyl) methanol, as studied by microwave spectroscopy and quantum chemical calculations. J. Phys. Chem. A 2006, 110 (18), 6054-6059.