Thermo Scientific Chemicals

2-Hydroxybenzophenone, 99%, Thermo Scientific Chemicals

Catalog number: L11100.06
5 g, Each
Thermo Scientific Chemicals

2-Hydroxybenzophenone, 99%, Thermo Scientific Chemicals

Catalog number: L11100.06
5 g, Each
Quantity
Catalog number: L11100.06
also known as L11100-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
117-99-7
IUPAC Name
2-benzoylphenol
Molecular Formula
C13H10O2
InChI Key
HJIAMFHSAAEUKR-UHFFFAOYSA-N
SMILES
OC1=CC=CC=C1C(=O)C1=CC=CC=C1
Appearance (Color)
Yellow
Identification (FTIR)
Conforms
Melting Point (clear melt)
36.0-42.0?C
Form
Crystals or powder or crystalline powder
Assay (GC)
≥98.5%

Description

2-Hydroxybenzophenone was used to quantify the metabolites of unmethylated extracts from the supernatants of Pseudomonas acidovorans cultures grown on 1,1-diphenylethylene. It reacts with 3-aminophenyl boronic acid to form macrocyclic boronates. 2-Hydroxybenzophenone forms copolymer 2-hydroxy, 3-allyl, 4,4?-dimethoxybenzophenone with methyl methacrylate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Hydroxybenzophenone was used to quantify the metabolites of unmethylated extracts from the supernatants of Pseudomonas acidovorans cultures grown on 1,1-diphenylethylene. It reacts with 3-aminophenyl boronic acid to form macrocyclic boronates. 2-Hydroxybenzophenone forms copolymer 2-hydroxy, 3-allyl, 4,4′-dimethoxybenzophenone with methyl methacrylate.

Solubility
Insoluble in water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text