2-Hydroxybenzophenone was used to quantify the metabolites of unmethylated extracts from the supernatants of Pseudomonas acidovorans cultures grown on 1,1-diphenylethylene. It reacts with 3-aminophenyl boronic acid to form macrocyclic boronates. 2-Hydroxybenzophenone forms copolymer 2-hydroxy, 3-allyl, 4,4?-dimethoxybenzophenone with methyl methacrylate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2-Hydroxybenzophenone was used to quantify the metabolites of unmethylated extracts from the supernatants of Pseudomonas acidovorans cultures grown on 1,1-diphenylethylene. It reacts with 3-aminophenyl boronic acid to form macrocyclic boronates. 2-Hydroxybenzophenone forms copolymer 2-hydroxy, 3-allyl, 4,4′-dimethoxybenzophenone with methyl methacrylate.
Solubility
Insoluble in water.
Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only
General References:
- A G Hay; D D Focht. Cometabolism of 1,1-dichloro-2,2-bis(4-chlorophenyl)ethylene by Pseudomonas acidovorans M3GY grown on biphenyl. Applied and Environmental Microbiology. 1998, 64, (6), 2141-2146.
- Charles Merritt, Gary W. Scott; Amitava Gupta; Andre Yavrouian. Transient absorption spectra of 2-hydroxybenzophenone photostabilizers. Chemical Physics Letters. 1980, 69, (1), 169-173.