Diallyl disulfide, tech. 80%, remainder mainly diallyl sulfide and diallyl trisulfide, Thermo Scientific Chemicals
Diallyl disulfide, tech. 80%, remainder mainly diallyl sulfide and diallyl trisulfide, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Diallyl disulfide, tech. 80%, remainder mainly diallyl sulfide and diallyl trisulfide, Thermo Scientific Chemicals

Diallyl disulfide, 80%, C6H10S2, CAS Number-2179-57-9, diallyldisulfide, allyl disulphide, 4,5-dithia-1,7-octadiene, 3-allyldisulfanyl-propene, 2-propenyl disulphide, diallyl disulfide, allyl disulfide, diallyl disulphide, disulfide, di-2-propenyl, 1,2-diallyldisulfane, 100g
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number L11106.14
also known as L11106-14
Price (USD)/ Each
51.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
60.50 
Save 8.85 (15%)
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
51.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
60.50 
Save 8.85 (15%)
Add to cart
Diallyl disulfide, tech. 80%, remainder mainly diallyl sulfide and diallyl trisulfide, Thermo Scientific Chemicals
Catalog numberL11106.14
Price (USD)/ Each
51.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
60.50 
Save 8.85 (15%)
-
Add to cart
Chemical Identifiers
CAS2179-57-9
IUPAC Name3-(prop-2-en-1-yldisulfanyl)prop-1-ene
Molecular FormulaC6H10S2
InChI KeyPFRGXCVKLLPLIP-UHFFFAOYSA-N
SMILESC=CCSSCC=C
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥78.0%
CommentContains 7-12% monosulfide and 3-7% trisulfide.
Identification (FTIR)Conforms
Refractive Index1.5370-1.5450 @ 20?C
Appearance (Color)Clear colorless to yellow
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only