(R)-(-)-Prolinol, 98+%, Thermo Scientific Chemicals
(R)-(-)-Prolinol, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

(R)-(-)-Prolinol, 98+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Catalog number L11109.03
also known as L11109-03
Price (USD)/ Each
78.60
-
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)/ Each
78.60
Add to cart
(R)-(-)-Prolinol, 98+%, Thermo Scientific Chemicals
Catalog numberL11109.03
Price (USD)/ Each
78.60
-
Add to cart
Chemical Identifiers
CAS68832-13-3
IUPAC Name(2R)-2-(hydroxymethyl)pyrrolidin-1-ium
Molecular FormulaC5H12NO
InChI KeyHVVNJUAVDAZWCB-RXMQYKEDSA-O
SMILESOC[C@H]1CCC[NH2+]1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Optical Rotation-32.0 ± 1.5? (c=1 in toluene)
Refractive Index1.4835-1.4875 @ 20?C
FormLiquid or viscous liquid
Assay (GC)≥98.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only