6-Chlorohexanoyl chloride, 95%, Thermo Scientific Chemicals
6-Chlorohexanoyl chloride, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

6-Chlorohexanoyl chloride, 95%, Thermo Scientific Chemicals

Have Questions?
Quantity:
5 g
Catalog number L11111.06
also known as L11111-06
Price (USD)/ Each
412.00
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
412.00
Add to cart
6-Chlorohexanoyl chloride, 95%, Thermo Scientific Chemicals
Catalog numberL11111.06
Price (USD)/ Each
412.00
-
Add to cart
Chemical Identifiers
CAS19347-73-0
IUPAC Name6-chlorohexanoyl chloride
Molecular FormulaC6H10Cl2O
InChI KeyWZILXAPNPKMOSA-UHFFFAOYSA-N
SMILESClCCCCCC(Cl)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Refractive Index1.4620-1.4680
Appearance (Color)Clear, colourless to pale brown
Formliquid
Assay (Titration ex Chloride)>94.0%
Assay (GC)>94.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only