Thermo Scientific Chemicals

Glycidyl methacrylate, 97%, stab. with 100ppm 4-methoxyphenol, Thermo Scientific Chemicals

Catalog number: L11133.18
50 g, Each
Thermo Scientific Chemicals

Glycidyl methacrylate, 97%, stab. with 100ppm 4-methoxyphenol, Thermo Scientific Chemicals

Catalog number: L11133.18
50 g, Each
Quantity
Catalog number: L11133.18
also known as L11133-18
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
106-91-2
IUPAC Name
(oxiran-2-yl)methyl 2-methylprop-2-enoate
Molecular Formula
C7H10O3
InChI Key
VOZRXNHHFUQHIL-UHFFFAOYNA-N
SMILES
CC(=C)C(=O)OCC1CO1
Appearance (Color)
Clear colorless to pale yellow
Assay (GC)
≥96.0%
Solution Test
(10% v/v in diethyl ether) : Clear (UK sourced material only)
Solution Test
(10% v/v in methanol) : Clear (UK sourced material only)
Stabilizer
4-Methoxyphenol

Description

Glycidyl methacrylate is commonly used as a monomer for the preparation of epoxy resins. It is also used to provide epoxy functionalization to polyolefins and other acrylate resins. Further, it is also useful as an epoxy resin additive for paint coating formulations and adhesive applications.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Glycidyl methacrylate is commonly used as a monomer for the preparation of epoxy resins. It is also used to provide epoxy functionalization to polyolefins and other acrylate resins. Further, it is also useful as an epoxy resin additive for paint coating formulations and adhesive applications.

Solubility
Miscible with benzene, alcohol and ether. Slightly miscible with water.

Notes
Light sensitive. Store in a cool place. Incompatible with strong oxidizing agents, strong acids and strong bases and peroxides.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text