Thermo Scientific Chemicals

(S)-(-)-1-Phenylethyl isocyanate, 98%, Thermo Scientific Chemicals

Catalog number: L11141.03
1 g, Each
Thermo Scientific Chemicals

(S)-(-)-1-Phenylethyl isocyanate, 98%, Thermo Scientific Chemicals

Catalog number: L11141.03
1 g, Each
Quantity
Catalog number: L11141.03
also known as L11141-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
14649-03-7
IUPAC Name
(1-isocyanatoethyl)benzene
Molecular Formula
C9H9NO
InChI Key
JJSCUXAFAJEQGB-UHFFFAOYNA-N
SMILES
CC(N=C=O)C1=CC=CC=C1
Appearance (Color)
Colorless to pale yellow
Form
Liquid
Assay (GC)
≥97.5%
Proton NMR
Conforms

Description

(S)-(-)-1-Phenylethyl isocyanate act by inhibition of cell proliferation and induction of apoptosis. It helps to prevent lung cancer and esophageal cancer. It is a fine chemical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(S)-(-)-1-Phenylethyl isocyanate act by inhibition of cell proliferation and induction of apoptosis. It helps to prevent lung cancer and esophageal cancer. It is a fine chemical intermediate.

Solubility
Hydrolyzes in water.

Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers. Containers which are opened must be carefully resealed and kept upright to prevent leakage. It is sensitive to moisture. Recommended storage temperature is 2 - 8°C.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text