Dimethyl cis-stilbene-4,4'-dicarboxylate, Thermo Scientific Chemicals
Dimethyl cis-stilbene-4,4'-dicarboxylate, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Dimethyl cis-stilbene-4,4'-dicarboxylate, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
250 mg
Catalog number L11142.03
also known as L11142-03
Price (USD)/ Each
106.00
-
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)/ Each
106.00
Add to cart
Dimethyl cis-stilbene-4,4'-dicarboxylate, Thermo Scientific Chemicals
Catalog numberL11142.03
Price (USD)/ Each
106.00
-
Add to cart
Chemical Identifiers
CAS143130-82-9
IUPAC Namemethyl 4-{2-[4-(methoxycarbonyl)phenyl]ethenyl}benzoate
Molecular FormulaC18H16O4
InChI KeyJOODVYOWCWQPMV-UHFFFAOYSA-N
SMILESCOC(=O)C1=CC=C(C=CC2=CC=C(C=C2)C(=O)OC)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale yellow
FormCrystals or powder or crystalline powder
Assay (HPLC)≥93.5%
Melting Point (clear melt)108.0-114.0?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only