Thermo Scientific Chemicals

Dimethyl cis-stilbene-4,4'-dicarboxylate, Thermo Scientific Chemicals

Catalog number: L11142.MD
250 mg, Each
Thermo Scientific Chemicals

Dimethyl cis-stilbene-4,4'-dicarboxylate, Thermo Scientific Chemicals

Catalog number: L11142.MD
250 mg, Each
Quantity
Catalog number: L11142.MD
also known as L11142-MD
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
143130-82-9
IUPAC Name
methyl 4-{2-[4-(methoxycarbonyl)phenyl]ethenyl}benzoate
Molecular Formula
C18H16O4
InChI Key
JOODVYOWCWQPMV-UHFFFAOYSA-N
SMILES
COC(=O)C1=CC=C(C=CC2=CC=C(C=C2)C(=O)OC)C=C1
Appearance (Color)
White to pale yellow
Form
Crystals or powder or crystalline powder
Assay (HPLC)
≥93.5%
Melting Point (clear melt)
108.0-114.0?C

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text