Thermo Scientific Chemicals

1,1,1-Trifluoro-2-propanol, 97%, Thermo Scientific Chemicals

Catalog number: L11150.14
25 g, Each
Thermo Scientific Chemicals

1,1,1-Trifluoro-2-propanol, 97%, Thermo Scientific Chemicals

Catalog number: L11150.14
25 g, Each
Quantity
Catalog number: L11150.14
also known as L11150-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
374-01-6
IUPAC Name
1,1,1-trifluoropropan-2-ol
Molecular Formula
C3H5F3O
InChI Key
GILIYJDBJZWGBG-UHFFFAOYNA-N
SMILES
CC(O)C(F)(F)F
Appearance (Color)
Clear colorless
Form
Liquid
Assay (GC)
≥96.0%
Comment
Specification differs for U.S. and non-U.S. material where indicated
Identification (FTIR)
Conforms

Description

1,1,1-Trifluoro-2-propanol is used as a chemical building block.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,1,1-Trifluoro-2-propanol is used as a chemical building block.

Solubility
Fully miscible in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with bases and oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text