Thermo Scientific Chemicals

2-(Trifluoromethyl)phenyl isocyanate, 97%, Thermo Scientific Chemicals

Catalog number: L11156.03
1 g, Each
Thermo Scientific Chemicals

2-(Trifluoromethyl)phenyl isocyanate, 97%, Thermo Scientific Chemicals

Catalog number: L11156.03
1 g, Each
Quantity
Catalog number: L11156.03
also known as L11156-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
2285-12-3
IUPAC Name
1-isocyanato-2-(trifluoromethyl)benzene
Molecular Formula
C8H4F3NO
InChI Key
GZWGTVZRRFPVAS-UHFFFAOYSA-N
SMILES
FC(F)(F)C1=CC=CC=C1N=C=O
Appearance (Color)
Clear colorless to pale yellow
Form
Liquid
Assay (GC)
≥96.0%
Comment
Specification differs for U.S. and non-U.S. material where indicated
Refractive Index
1.4700-1.4760 @ 20°C (non-U.S. specification)

Description

2-(Trifluoromethyl)phenyl isocyanate is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-(Trifluoromethyl)phenyl isocyanate is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields.

Solubility
Hydrolyzes in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with bases and oxidizing agents. It is sensitive to moisture. Store under dry inert gas.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text