2.5% w/v solution in acetone will be transparent to almost transparent by JIS K8001 5.2
Melting Point
104-112?C (with ≤3? range)
Form
crystalline powder
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.