1,2-Benzenedithiol is used in preparation of schiff base, which is employed to make new series of copper(II) and zinc(II) complexes.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
1,2-Benzenedithiol is used in preparation of schiff base, which is employed to make new series of copper(II) and zinc(II) complexes.
Solubility
Soluble in water, benzene, ethanol, diethyl ether, ethyl acetate.
Notes
Incompatible with bases and strong oxidizing agents.
RUO – Research Use Only
General References:
- Giguere, J.; Morin, J. Superextended Tetrathiafulvalene: Synthesis, Optoelectronic Properties, Fullerenes Complexation, and Photooxidation Study. J. Org. Chem. 2015, 80 (13), 6767-6775.
- Nouchi, R.; Tanimoto, T. Substituent-Controlled Reversible Switching of Charge Injection Barrier Heights at Metal/Organic Semiconductor Contacts Modified with Disordered Molecular Monolayers. ACS Nano. 2015, 9 (7), 7429-7439.