Thermo Scientific Chemicals

5-Acetamido-2-aminobenzoic acid, 97%, Thermo Scientific Chemicals

Catalog number: L11256.06
5 g, Each
Thermo Scientific Chemicals

5-Acetamido-2-aminobenzoic acid, 97%, Thermo Scientific Chemicals

Catalog number: L11256.06
5 g, Each
Quantity
Catalog number: L11256.06
also known as L11256-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
50670-83-2
IUPAC Name
2-amino-5-acetamidobenzoic acid
Molecular Formula
C9H10N2O3
InChI Key
GSOHXJQXAKNJES-UHFFFAOYSA-N
SMILES
CC(=O)NC1=CC=C(N)C(=C1)C(O)=O
Form
Powder
Appearance (Color)
White to grey to brown
Assay (HPLC)
≥96.0%
Assay (Aqueous acid-base Titration)
≥96.0 to ≤104.0%

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text