Lectin phytohemagglutinin-L (PHA-L) is strongly inhibited by N-acetylglucosamine β(1-2) mannopyranosyl residues. This lectin has been shown to agglutinate leukocytes andRead more
Have Questions?
Catalog Number
Quantity
L11270
1 mg
Catalog number L11270
Price (KRW)
272,000
Online offer
Ends: 30-Jun-2025
319,000
Save 47,000 (15%)
Each
-
Add to cart
Quantity:
1 mg
Price (KRW)
272,000
Online offer
Ends: 30-Jun-2025
319,000
Save 47,000 (15%)
Each
Add to cart
Ask our AI about this Product
Lectin phytohemagglutinin-L (PHA-L) is strongly inhibited by N-acetylglucosamine β(1-2) mannopyranosyl residues. This lectin has been shown to agglutinate leukocytes and induce mitogenic activity in T-lymphocytes and has also been used as an anterograde tracer for the study of neuronal pathways.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Label TypeAlexa Fluor Dyes
Product LineAlexa Fluor™
Protein SubtypeLectins
Quantity1 mg
Shipping ConditionRoom Temperature
SourceRed kidney bean
ConjugateAlexa Fluor 488
FormLyophilized
Unit SizeEach
Contents & Storage
Store in freezer (-5 to -30°C) and protect from light.
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our
Privacy Notice.