4-Heptyn-2-ol, 97%, Thermo Scientific Chemicals
4-Heptyn-2-ol, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4-Heptyn-2-ol, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Catalog number L11292.14
also known as L11292-14
Price (USD)/ Each
191.00
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
191.00
Add to cart
4-Heptyn-2-ol, 97%, Thermo Scientific Chemicals
Catalog numberL11292.14
Price (USD)/ Each
191.00
-
Add to cart
Chemical Identifiers
CAS19781-81-8
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear, colourless to pale yellow
Formliquid
Assay (GC)> 96.0%
Refractive Index1.4490 - 1.4540 @20?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only