N-(2-Hydroxyethyl)succinimide, 95%, Thermo Scientific Chemicals
N-(2-Hydroxyethyl)succinimide, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

N-(2-Hydroxyethyl)succinimide, 95%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Catalog number L11306.14
also known as L11306-14
Price (USD)/ Each
123.00
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
123.00
Add to cart
N-(2-Hydroxyethyl)succinimide, 95%, Thermo Scientific Chemicals
Catalog numberL11306.14
Price (USD)/ Each
123.00
-
Add to cart
Chemical Identifiers
CAS18190-44-8
IUPAC Name1-(2-hydroxyethyl)pyrrolidine-2,5-dione
Molecular FormulaC6H9NO3
InChI KeyTWYIPMITVXPNEM-UHFFFAOYSA-N
SMILESOCCN1C(=O)CCC1=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream to pale yellow
Assay (HPLC)≥94.0%
FormCrystals or powder or crystalline powder or lumps or fused solid
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only