Thermo Scientific Chemicals

1,1,1-Trifluoroacetone cyanohydrin, 95%, Thermo Scientific Chemicals

Catalog number: L11318.06
5 g, Each
Thermo Scientific Chemicals

1,1,1-Trifluoroacetone cyanohydrin, 95%, Thermo Scientific Chemicals

Catalog number: L11318.06
5 g, Each
Quantity
Catalog number: L11318.06
also known as L11318-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
335-08-0
IUPAC Name
3,3,3-trifluoro-2-hydroxy-2-methylpropanenitrile
Molecular Formula
C4H4F3NO
InChI Key
XDCMNDCKYSQKAX-UHFFFAOYNA-N
SMILES
CC(O)(C#N)C(F)(F)F
Refractive Index
1.3350-1.3410 @ 20?C
Form
Liquid
Assay (unspecified)
(by Proton NMR) : >94.0%
Appearance (Color)
Clear, colorless

Description

A chlorinated acetone derivative for organic synthesis and proteomics research

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
A chlorinated acetone derivative for organic synthesis and proteomics research

Solubility
Fully miscible in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents. It is sensitive to moisture. Store under dry inert gas.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text