Thermo Scientific Chemicals

5,7-Dimethylisatin, 96%, Thermo Scientific Chemicals

Catalog number: L11332.14
25 g, Each
Thermo Scientific Chemicals

5,7-Dimethylisatin, 96%, Thermo Scientific Chemicals

Catalog number: L11332.14
25 g, Each
Quantity
Catalog number: L11332.14
also known as L11332-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
39603-24-2
IUPAC Name
5,7-dimethyl-2,3-dihydro-1H-indole-2,3-dione
Molecular Formula
C10H9NO2
InChI Key
HFZSCCJTJGWTDZ-UHFFFAOYSA-N
SMILES
CC1=CC(C)=C2NC(=O)C(=O)C2=C1
Assay (HPLC)
≥95.0%
Appearance (Color)
Orange to dark red to red to dark brown to brown
Form
Powder

Description

5,7-Dimethylisatin is used to produce cyano-(5,7-dimethyl-2-oxo-1,2-dihydro-indol-3-ylidene)-acetic acid ethyl ester by knoevenagel condensation with cyanoacetic acid ethyl ester. It is an important raw material and intermediate used in organic synthesis, pharmaceuticals and agrochemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5,7-Dimethylisatin is used to produce cyano-(5,7-dimethyl-2-oxo-1,2-dihydro-indol-3-ylidene)-acetic acid ethyl ester by knoevenagel condensation with cyanoacetic acid ethyl ester. It is an important raw material and intermediate used in organic synthesis, pharmaceuticals and agrochemicals.

Solubility
Insoluble in water.

Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers. Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text