Thermo Scientific Chemicals

Benzo[b]thiophene-2-carboxylic hydrazide, 97%, Thermo Scientific Chemicals

Catalog number: L11357.06
5 g, Each
Thermo Scientific Chemicals

Benzo[b]thiophene-2-carboxylic hydrazide, 97%, Thermo Scientific Chemicals

Catalog number: L11357.06
5 g, Each
Quantity
Catalog number: L11357.06
also known as L11357-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
175135-07-6
IUPAC Name
1-benzothiophene-2-carbohydrazide
Molecular Formula
C9H8N2OS
InChI Key
ZXKPFIRPUUAAPQ-UHFFFAOYSA-N
SMILES
NNC(=O)C1=CC2=CC=CC=C2S1
Appearance (Color)
White to cream to orange or pale pink
Assay (HPLC)
≥96.0%
Form
Crystals or powder or crystalline powder
Melting Point (clear melt)
183.5-189.5?C

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text