Thermo Scientific Chemicals

1-Phenylisatin, 98%, Thermo Scientific Chemicals

Catalog number: L11388.06
5 g, Each
Thermo Scientific Chemicals

1-Phenylisatin, 98%, Thermo Scientific Chemicals

Catalog number: L11388.06
5 g, Each
Quantity
Catalog number: L11388.06
also known as L11388-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
723-89-7
Appearance (Color)
Dark orange to orange to red
Assay (HPLC)
≥97.5%
Form
Crystals or powder or crystalline powder or fused solid
Melting Point (clear melt)
137.0-143.0?C

Description

1-Phenylisatin is a reactant for preparation of indolin-2,3-dione Schiff bases as potential antimycobacterial agents, antipyretic agents, antiproliferative agents, phenylacetamides as anticonvulsant agents and selective Galanin GAL3 receptor antagonists.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Phenylisatin is a reactant for preparation of indolin-2,3-dione Schiff bases as potential antimycobacterial agents, antipyretic agents, antiproliferative agents, phenylacetamides as anticonvulsant agents and selective Galanin GAL3 receptor antagonists.

Solubility
Insoluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text