Thermo Scientific Chemicals

3,3'-Diaminodiphenyl sulfone, 98%, Thermo Scientific Chemicals

Catalog number: L11400.14
25 g, Each
Thermo Scientific Chemicals

3,3'-Diaminodiphenyl sulfone, 98%, Thermo Scientific Chemicals

Catalog number: L11400.14
25 g, Each
Quantity
Catalog number: L11400.14
also known as L11400-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
599-61-1
Assay (Non-aqueous acid-base Titration)
≥97.5 to ≤102.5%
Assay (HPLC)
≥97.5%
Appearance (Color)
White to cream to orange
Form
Crystals or powder or crystalline powder
Melting Point (clear melt)
170.0-177.0?C

Description

3,3'-Diaminodiphenyl sulfone is used for curing agent for epoxy resin and also important monomer for polysulfonamide fiber(PSA).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3,3′-Diaminodiphenyl sulfone is used for curing agent for epoxy resin and also important monomer for polysulfonamide fiber(PSA).

Solubility
Insoluble in water.

Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers. Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text