1-Thio-beta-D-glucose tetraacetate, 98+%, Thermo Scientific Chemicals
1-Thio-beta-D-glucose tetraacetate, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

1-Thio-beta-D-glucose tetraacetate, 98+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
250 mg
Catalog number L11404.03
also known as L11404-03
Price (USD)/ Each
90.65
Online exclusive
101.00 
Save 10.35 (10%)
-
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)/ Each
90.65
Online exclusive
101.00 
Save 10.35 (10%)
Add to cart
1-Thio-beta-D-glucose tetraacetate, 98+%, Thermo Scientific Chemicals
Catalog numberL11404.03
Price (USD)/ Each
90.65
Online exclusive
101.00 
Save 10.35 (10%)
-
Add to cart
Chemical Identifiers
CAS19879-84-6
IUPAC Name[3,4,5-tris(acetyloxy)-6-sulfanyloxan-2-yl]methyl acetate
Molecular FormulaC14H20O9S
InChI KeySFOZKJGZNOBSHF-UHFFFAOYNA-N
SMILESCC(=O)OCC1OC(S)C(OC(C)=O)C(OC(C)=O)C1OC(C)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (Argentometric Titration)≥97.0 to ≤102.0%
Identification (FTIR)Conforms
Appearance (Color)White
FormCrystals or powder or crystalline powder
Melting Point (clear melt)113.0-119.0°C
View more
1-Thio-beta-D-glucose tetraacetate is used as an inhibitor of the Maillard reaction between glucose and glycine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Thio-beta-D-glucose tetraacetate is used as an inhibitor of the Maillard reaction between glucose and glycine.

Solubility
Soluble in water and chloroform (50 mg/ml).

Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers. Incompatible with strong oxidizing agents.
RUO – Research Use Only

General References:

  1. J. Gálvez.; R. Garc.; Domenech.; M.J. Gómez-Lechón.; J.V. Castellb. Use of molecular topology in the selection of new cytostatic drugs. Tetrahedron Lett. 2000, 504 (1-3), 241-248.
  2. Welling, Mick M..; Alberto.; Roger. Performance of a 99mTc-labelled 1-thio-β-D-glucose 2,3,4,6-tetra-acetate analogue in the detection of infections and tumours in mice: a comparison with [18F]FDG. Nuclear Medicine Communications. 2010, 31 (3), 239-248.