Thermo Scientific Chemicals

1-Thio-beta-D-glucose tetraacetate, 98+%, Thermo Scientific Chemicals

Catalog number: L11404.MD
250 mg, Each
Thermo Scientific Chemicals

1-Thio-beta-D-glucose tetraacetate, 98+%, Thermo Scientific Chemicals

Catalog number: L11404.MD
250 mg, Each
Quantity
Catalog number: L11404.MD
also known as L11404-MD
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
19879-84-6
IUPAC Name
[3,4,5-tris(acetyloxy)-6-sulfanyloxan-2-yl]methyl acetate
Molecular Formula
C14H20O9S
InChI Key
SFOZKJGZNOBSHF-UHFFFAOYNA-N
SMILES
CC(=O)OCC1OC(S)C(OC(C)=O)C(OC(C)=O)C1OC(C)=O
Appearance (Color)
White
Form
Crystals or powder or crystalline powder
Assay (Argentometric Titration)
≥97.0 to ≤102.0%
Identification (FTIR)
Conforms
Melting Point (clear melt)
113.0-119.0°C

Description

1-Thio-beta-D-glucose tetraacetate is used as an inhibitor of the Maillard reaction between glucose and glycine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Thio-beta-D-glucose tetraacetate is used as an inhibitor of the Maillard reaction between glucose and glycine.

Solubility
Soluble in water and chloroform (50 mg/ml).

Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers. Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text