N-Benzylacrylamide, 96%, Thermo Scientific Chemicals
N-Benzylacrylamide, 96%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

N-Benzylacrylamide, 96%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Catalog number L11450.06
also known as L11450-06
Price (USD)/ Each
164.00
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
164.00
Add to cart
N-Benzylacrylamide, 96%, Thermo Scientific Chemicals
Catalog numberL11450.06
Price (USD)/ Each
164.00
-
Add to cart
Chemical Identifiers
CAS13304-62-6
IUPAC NameN-benzylprop-2-enamide
Molecular FormulaC10H11NO
InChI KeyOHLHOLGYGRKZMU-UHFFFAOYSA-N
SMILESC=CC(=O)NCC1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
FormPowder
Melting Point (clear melt)57.5-66.5?C
Appearance (Color)White to yellow to brown
Assay (GC)≥95.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only