Thermo Scientific Chemicals

4-Hydroxy-2-butanone, 95%, Thermo Scientific Chemicals

Catalog number: L11456.14
25 g, Each
Thermo Scientific Chemicals

4-Hydroxy-2-butanone, 95%, Thermo Scientific Chemicals

Catalog number: L11456.14
25 g, Each
Quantity
Catalog number: L11456.14
also known as L11456-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
590-90-9
IUPAC Name
4-hydroxybutan-2-one
Molecular Formula
C4H8O2
InChI Key
LVSQXDHWDCMMRJ-UHFFFAOYSA-N
SMILES
CC(=O)CCO
Form
Liquid
Appearance (Color)
Clear colorless to yellow
Assay (GC)
≥94.0%
Refractive Index
1.4265-1.4325 @ 20°C (non-U.S. specification)
Identification (FTIR)
Conforms (non-U.S. specification)

Description

4-Hydroxy-2-butanone is used in the preparation of fused benzazepine molecules as selective D3 receptor antagonists with pharmaceutical activity. It is also used in the preparation of verrucarin and mevalonic acid lactone. Further, it is used to prepare 3-buten-2-one through dehydration over an anatase titanium dioxide catalyst. It is also employed in the preparation of 4-methyl-5-hydroxymethylthiazole, (±)-lineatin and beta-hydroxylactones.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Hydroxy-2-butanone is used in the preparation of fused benzazepine molecules as selective D3 receptor antagonists with pharmaceutical activity. It is also used in the preparation of verrucarin and mevalonic acid lactone. Further, it is used to prepare 3-buten-2-one through dehydration over an anatase titanium dioxide catalyst. It is also employed in the preparation of 4-methyl-5-hydroxymethylthiazole, (±)-lineatin and beta-hydroxylactones.

Solubility
Miscible with water, alcohol, ethanol and ether.

Notes
Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text