Benzoyl fluoride, 97%, Thermo Scientific Chemicals
Benzoyl fluoride, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Benzoyl fluoride, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number L11464.06
also known as L11464-06
Price (USD)/ Each
33.65
Online exclusive
36.80 
Save 3.15 (9%)
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
33.65
Online exclusive
36.80 
Save 3.15 (9%)
Add to cart
Benzoyl fluoride, 97%, Thermo Scientific Chemicals
Catalog numberL11464.06
Price (USD)/ Each
33.65
Online exclusive
36.80 
Save 3.15 (9%)
-
Add to cart
Chemical Identifiers
CAS455-32-3
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow to pale brown
FormLiquid
Assay (GC)≥96.0%
Identification (FTIR)Conforms
Refractive Index1.4930-1.4990 @ 20°C
Benzoyl fluoride is used as a mild fluorinating agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Benzoyl fluoride is used as a mild fluorinating agent.

Solubility
Hydrolyses in water.

Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers. It is sensitive to moisture. Store at room temperature.
RUO – Research Use Only

General References:

  1. Byeong Doo Song.; William P. Jencks. Aminolysis of benzoyl fluorides in water. J. Am. Chem. Soc. 1989, 111 (22), 8479-8484.
  2. Timo Asikainen.; Mikko Ritala.; Wei-Min Li.; Reijo Lappalainen.; Markku Leskelä. Modifying ALE grown In2O3 films by benzoyl fluoride pulses. Applied Surface Science. 1997, 112 231-235.