Dimethyl D-tartrate, 99%, Thermo Scientific Chemicals
Dimethyl D-tartrate, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Dimethyl D-tartrate, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number L11472.06
also known as L11472-06
Price (USD)/ Each
56.90
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
56.90
Add to cart
Dimethyl D-tartrate, 99%, Thermo Scientific Chemicals
Catalog numberL11472.06
Price (USD)/ Each
56.90
-
Add to cart
Chemical Identifiers
CAS13171-64-7
IUPAC Name1,4-dimethyl (2S,3S)-2,3-dihydroxybutanedioate
Molecular FormulaC6H10O6
InChI KeyPVRATXCXJDHJJN-IMJSIDKUSA-N
SMILESCOC(=O)[C@@H](O)[C@H](O)C(=O)OC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
FormCrystals or powder or crystalline powder or lumps or fused solid
Optical Rotation-21.7 ± 0.1? (C=1 in water)
Assay (GC)≥98.5%
Refractive Index1.4525-1.4555 @ 20?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only