4-Ethoxyphenyl isocyanate, 97%, Thermo Scientific Chemicals
4-Ethoxyphenyl isocyanate, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4-Ethoxyphenyl isocyanate, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Catalog number L11509.14
also known as L11509-14
Price (USD)/ Each
127.00
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
127.00
Add to cart
4-Ethoxyphenyl isocyanate, 97%, Thermo Scientific Chemicals
Catalog numberL11509.14
Price (USD)/ Each
127.00
-
Add to cart
Chemical Identifiers
CAS32459-62-4
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥96.0%
Appearance (Color)Clear colorless to yellow
Refractive Index1.5330-1.5380 @ 20?C
FormLiquid
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only