Thermo Scientific Chemicals

Bis(2-bromoethyl) ether, 95%, Thermo Scientific Chemicals

Catalog number: L11510.14
25 g, Each
Thermo Scientific Chemicals

Bis(2-bromoethyl) ether, 95%, Thermo Scientific Chemicals

Catalog number: L11510.14
25 g, Each
Quantity
Catalog number: L11510.14
also known as L11510-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
5414-19-7
IUPAC Name
1-bromo-2-(2-bromoethoxy)ethane
Molecular Formula
C4H8Br2O
InChI Key
FOZVXADQAHVUSV-UHFFFAOYSA-N
SMILES
BrCCOCCBr
Form
Liquid
Refractive Index
1.5110-1.5170 @ 20?C
Identification (FTIR)
Conforms
Appearance (Color)
Clear colorless to yellow to brown
Assay (GC)
≥94.0%

Description

Bis(2-bromoethyl) ether is used dfor synthesis of heterocyclic and crown ether. It is also used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Bis(2-bromoethyl) ether is used dfor synthesis of heterocyclic and crown ether. It is also used as a pharmaceutical intermediate.

Solubility
Not miscible or difficult to mix with water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents and bases.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text