Thermo Scientific Chemicals

3-Hydroxyphenylacetylene, 97%, Thermo Scientific Chemicals

Catalog number: L11523.06
5 g, Each
Thermo Scientific Chemicals

3-Hydroxyphenylacetylene, 97%, Thermo Scientific Chemicals

Catalog number: L11523.06
5 g, Each
Quantity
Catalog number: L11523.06
also known as L11523-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
10401-11-3
IUPAC Name
3-ethynylphenol
Molecular Formula
C8H6O
InChI Key
AODMJIOEGCBUQL-UHFFFAOYSA-N
SMILES
OC1=CC=CC(=C1)C#C
Assay (GC)
>96.0%
Appearance (Color)
Clear colorless to yellow or brown
Form
Liquid
Refractive Index
1.5845-1.5895 @ 20?C

Description

3-Hydroxyphenylacetylene is used as a fluorogenic and chromogenic probe for bacterial enzymes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Hydroxyphenylacetylene is used as a fluorogenic and chromogenic probe for bacterial enzymes.

Solubility
Miscible with dimethylformamide.

Notes
Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text